accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONCH3NClHNO

CAS RN 85-18-7

IUPAC Name: 8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 8-chlorotheophylline 茶氯酸异丙嗪 moxastine theoclate promethazine teoclate
Molecular Weight (g/mol): 214.61
Molecular Formula: C7H7ClN4O2
InChi Key: RYIGNEOBDRVTHA-UHFFFAOYSA-N
SMILES: CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results