accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHHO

CAS RN 85-27-8

IUPAC Name: 4-(1-phenylethyl)benzene-1,3-diol
Synonyms: phenylethyl resorcinol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: PQSXNIMHIHYFEE-UHFFFAOYNA-N
SMILES: CC(C1=CC=CC=C1)C1=C(O)C=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results