accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHO

CAS RN 85-39-2

IUPAC Name: [3-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Synonyms: [3-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: IGHHPVIMEQGKNE-UHFFFAOYNA-N
SMILES: OCC1C2CC(C=C2)C1CO
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results