accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 85-42-7

IUPAC Name: octahydro-2-benzofuran-1,3-dione
Synonyms: hexahydrophthalic anhydride HHPA
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: MUTGBJKUEZFXGO-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2CCCCC12
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)