accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 85-44-9

IUPAC Name: 1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: phthalic anhydride wiltrol P retarder AK retarder PD ESEN vulkalent B/C anidride ftalica Pan anhydride phtalique anhydrid kyseliny ftalove
Molecular Weight (g/mol): 148.12
Molecular Formula: C8H4O3
InChi Key: LGRFSURHDFAFJT-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (16)
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Boiling Point 
  • (7)
  • (7)
  • (3)
Percent Purity 
  • (1)
  • (4)
  • (5)
  • (5)
Grade 
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (4)
  • (6)
  • (1)
  • (16)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (7)
  • (7)
  • (3)
  • (1)
  • (4)
  • (5)
  • (5)
  • (6)
  • (3)
  • (1)
  • (5)
  • (1)