accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOO

CAS RN 85-46-1

IUPAC Name: naphthalene-1-sulfonyl chloride
Synonyms: 1-naphthalenesulfonyl chloride
Molecular Weight (g/mol): 226.67
Molecular Formula: C10H7ClO2S
InChi Key: DASJFYAPNPUBGG-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)