accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85-47-2

OHSOO

CAS RN 85-47-2

IUPAC Name: naphthalene-1-sulfonic acid
Synonyms: α-naphthalenesulfonic acid
Molecular Weight (g/mol): 208.23
Molecular Formula: C10H8O3S
InChi Key: PSZYNBSKGUBXEH-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results