accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOO

CAS RN 85-55-2

IUPAC Name: 2-(4-methylbenzoyl)benzoic acid
Synonyms: 2-(4-methylbenzoyl)benzoic acid
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: ICQOWIXIHDDXDI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results