accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85-68-7

IUPAC Name: 1-benzyl 2-butyl benzene-1,2-dicarboxylate
Synonyms: benzyl butyl phthalate butylbenzyl phthalate unimoll BB butyl benzyl phthalate (substance) benzyl butyl phthalate (bbp) BBP sicol 160 sicol caswell no. 125G ccris 104
Molecular Weight (g/mol): 312.37
Molecular Formula: C19H20O4
InChi Key: IRIAEXORFWYRCZ-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)