accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85-81-4

IUPAC Name: 6-methoxy-8-nitroquinoline
Synonyms: quinoline, 6-methoxy-8-nitro-
Molecular Weight (g/mol): 204.19
Molecular Formula: C10H8N2O3
InChi Key: MIMUSZHMZBJBPO-UHFFFAOYSA-N
SMILES: COC1=CC(=C2N=CC=CC2=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)