accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COClH3N

CAS RN 85006-21-9

IUPAC Name: 2-chloro-5-methoxyanilinium chloride
Synonyms: 2-chloro-5-methoxyanilinium chloride
Molecular Weight (g/mol): 194.06
Molecular Formula: C7H9Cl2NO
InChi Key: NQWBPXKJBZYGHZ-UHFFFAOYSA-N
SMILES: [Cl-].COC1=CC=C(Cl)C([NH3+])=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results