accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2BOHHO

CAS RN 85006-23-1

IUPAC Name: (3-aminophenyl)boronic acid hydrochloride
Synonyms: M-aminophenylboronic acid hydrochloride
Molecular Weight (g/mol): 173.4
Molecular Formula: C6H9BClNO2
InChi Key: QBMHZZHJIBUPOX-UHFFFAOYSA-N
SMILES: Cl.NC1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)