accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85006-31-1

IUPAC Name: methyl 3-amino-4-methylthiophene-2-carboxylate
Synonyms: methyl 3-amino-4-methylthiophene-2-carboxylate
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: YICRPERKKBDRSP-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C(C)=CS1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)