accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFFF

CAS RN 85013-98-5

IUPAC Name: 1-[4-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[4-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: MOEXTBIPPMLEFX-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OC(F)(F)F)C=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)