accessibility menu, dialog, popup

UserName

Viewing profile as user:
BaII

CAS RN 85017-89-6

IUPAC Name: barium(2+) diiodide
Synonyms: barium iodide
Molecular Weight (g/mol): 391.14
Molecular Formula: BaI2
InChi Key: SGUXGJPBTNFBAD-UHFFFAOYSA-L
SMILES: [I-].[I-].[Ba++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results