accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850349-02-9

BrNN

CAS RN 850349-02-9

IUPAC Name: 8-bromoimidazo[1,2-a]pyridine
Synonyms: 8-bromoimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: GCCVNNZARPFXES-UHFFFAOYSA-N
SMILES: BrC1=CC=CN2C=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results