CAS RN 850374-99-1
IUPAC Name:
5-bromo-1-benzothiophene-3-carbonyl chloride
Synonyms:
5-bromo-1-benzothiophene-3-carbonyl chloride
Molecular Weight (g/mol):
275.54
Molecular Formula:
C9H4BrClOS
InChi Key:
DNAFRKYZNZZCCO-UHFFFAOYSA-N
SMILES:
ClC(=O)C1=CSC2=CC=C(Br)C=C12
1
–
1
of
1
results
1
5-Bromobenzo[b]thiophene-3-carbonyl chloride, ≥97%, Thermo Scientific™
CAS: 850374-99-1 Molecular Formula: C9H4BrClOS Molecular Weight (g/mol): 275.54 MDL Number: MFCD07368522 InChI Key: DNAFRKYZNZZCCO-UHFFFAOYSA-N Synonym: 5-bromobenzo b thiophene-3-carbonyl chloride,benzo b thiophene-3-carbonylchloride, 5-bromo PubChem CID: 4961276 IUPAC Name: 5-bromo-1-benzothiophene-3-carbonyl chloride SMILES: ClC(=O)C1=CSC2=CC=C(Br)C=C12