accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNO

CAS RN 850375-08-5

IUPAC Name: tert-butyl 4-[(3-formylphenyl)methyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[(3-formylphenyl)methyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 304.39
Molecular Formula: C17H24N2O3
InChi Key: YIKZZJRCKPEHGU-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC2=CC=CC(C=O)=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results