accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSOBr

CAS RN 850375-12-1

IUPAC Name: 2-bromo-1-(5-bromo-1-benzothiophen-3-yl)ethan-1-one
Synonyms: 2-bromo-1-(5-bromo-1-benzothiophen-3-yl)ethanone
Molecular Weight (g/mol): 334.03
Molecular Formula: C10H6Br2OS
InChi Key: QFZPWQQHQRYUKB-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CSC2=CC=C(Br)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results