accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CONOOCH3

CAS RN 850411-14-2

IUPAC Name: ethyl 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperidine-3-carboxylate
Synonyms: ethyl 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperidine-3-carboxylate
Molecular Weight (g/mol): 387.28
Molecular Formula: C21H30BNO5
InChi Key: YQADGHATUTVIEA-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCN(C1)C(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results