accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850567-91-8

IUPAC Name: (4-tert-butylcyclohex-1-en-1-yl)boronic acid
Synonyms: 4-tert-butylcyclohex-1-en-1-ylboronic acid
Molecular Weight (g/mol): 182.07
Molecular Formula: C10H19BO2
InChi Key: QPJWLDFYBBQYSG-UHFFFAOYNA-N
SMILES: CC(C)(C)C1CCC(=CC1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results