accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850567-97-4

IUPAC Name: (2-methanesulfinylphenyl)boronic acid
Synonyms: 2-methanesulfinylphenylboronic acid
Molecular Weight (g/mol): 184.02
Molecular Formula: C7H9BO3S
InChi Key: PHORKVSBWZGTEX-UHFFFAOYNA-N
SMILES: CS(=O)C1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results