accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850568-66-0

IUPAC Name: [6-(benzyloxy)-1-[(tert-butoxy)carbonyl]-1H-indol-2-yl]boronic acid
Synonyms: 6-(benzyloxy)-1-(tert-butoxycarbonyl)indol-2-ylboronic acid
Molecular Weight (g/mol): 367.21
Molecular Formula: C20H22BNO5
InChi Key: OGONSJPAZMVGTG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C(=CC2=CC=C(OCC3=CC=CC=C3)C=C12)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results