accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFF

CAS RN 85068-27-5

IUPAC Name: 2-(2,5-difluorophenyl)acetate
Synonyms: 2-(2,5-difluorophenyl)acetate
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H5F2O2
InChi Key: FKCRTRYQHZHXES-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC(F)=CC=C1F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)