accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3FFFFFF

CAS RN 85068-29-7

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]methanaminium
Synonyms: [3,5-bis(trifluoromethyl)phenyl]methanaminium
Molecular Weight (g/mol): 244.16
Molecular Formula: C9H8F6N
InChi Key: DHVHORCFFOSRBP-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)