accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85070-47-9

IUPAC Name: 1-(bromomethyl)-3-chloro-2-fluorobenzene
Synonyms: 1-(bromomethyl)-3-chloro-2-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: HYILLTADABKYHO-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC=C1CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results