accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850881-09-3

IUPAC Name: 2-(3-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 294.26
Molecular Formula: C16H27BO2S
InChi Key: XCXAUPBHQCCWCI-UHFFFAOYSA-N
SMILES: CCCCCCC1=C(SC=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results