accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 851042-10-9

IUPAC Name: 9,9-dioctyl-9H-fluorene-2,7-diamine
Synonyms: 9,9-dioctylfluorene-2,7-diamine
Molecular Weight (g/mol): 420.69
Molecular Formula: C29H44N2
InChi Key: ODJBGRHFVFWTBO-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(N)=C2)C2=C1C=C(N)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)