accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 851051-43-9

H3CSOONNClRu

CAS RN 851051-43-9

IUPAC Name: chlororutheniumbis(ylium); {2-[(4-methylbenzenesulfonyl)azanidyl]-1,2-diphenylethyl}(3-phenylpropyl)azanide
Synonyms: chlororutheniumbis(ylium); {2-[(4-methylbenzenesulfonyl)azanidyl]-1,2-diphenylethyl}(3-phenylpropyl)azanide
Molecular Weight (g/mol): 619.16
Molecular Formula: C30H30ClN2O2RuS
InChi Key: XLHIJKCWYHFMEK-UHFFFAOYNA-M
SMILES: Cl[Ru++].CC1=CC=C(C=C1)S(=O)(=O)[N-]C(C([N-]CCCC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results