accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85107-55-7

IUPAC Name: [4-(methoxycarbonyl)-2-nitrophenyl]boronic acid
Synonyms: 4-(methoxycarbonyl)-2-nitrophenylboronic acid
Molecular Weight (g/mol): 224.96
Molecular Formula: C8H8BNO6
InChi Key: SFEJGHJCGQNFQC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(B(O)O)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results