accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFBr

CAS RN 85118-00-9

IUPAC Name: 2-(bromomethyl)-1,3-difluorobenzene
Synonyms: 2-(bromomethyl)-1,3-difluorobenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: LSXJPJGBWSZHTM-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1CBr
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)