accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHSOOBr

CAS RN 851215-65-1

IUPAC Name: 2-bromo-N-(naphthalen-1-yl)benzene-1-sulfonamide
Synonyms: 2-bromo-N-(naphthalen-1-yl)benzenesulfonamide
Molecular Weight (g/mol): 362.24
Molecular Formula: C16H12BrNO2S
InChi Key: LSOKWJIMCSVKBL-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1S(=O)(=O)NC1=CC=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results