accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOHFF

CAS RN 851386-31-7

IUPAC Name: 2,3-difluoropyridine-4-carboxylic acid
Synonyms: 2,3-difluoropyridine-4-carboxylic acid
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: FFGHMEKFPCGYEG-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)