accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85144-11-2

IUPAC Name: lithium(1+) chloride
Synonyms: lithium chloride|LiCl|lithiumchlorid|chlorure de lithium|cloruro de litio|lithii chloridum
Molecular Weight (g/mol): 42.39
Molecular Formula: ClLi
InChi Key: KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES: [Li+].[Cl-]
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)