accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85160-84-5

IUPAC Name: 2,2-dimethyl-6-nitro-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 2,2-dimethyl-6-nitro-4H-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 222.2
Molecular Formula: C10H10N2O4
InChi Key: YKXZRZGZJZYBBH-UHFFFAOYSA-N
SMILES: CC1(C)OC2=C(NC1=O)C=C(C=C2)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results