accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 851768-63-3

IUPAC Name: 5-amino-1',3'-dihydro-4H-spiro[naphthalene-1,2'-perimidin]-4-one
Synonyms: 5-amino-1',3'-dihydrospiro[naphthalene-1,2'-perimidin]-4-one
Molecular Weight (g/mol): 313.36
Molecular Formula: C20H15N3O
InChi Key: GEUHINMXSZXTFV-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1C(=O)C=CC21NC2=CC=CC3=CC=CC(N1)=C23
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results