accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 852138-89-7

IUPAC Name: 4,4'-dibromo-2,2'-diiodo-1,1'-biphenyl
Synonyms: 4,4'-dibromo-2,2'-diiodo-1,1'-biphenyl
Molecular Weight (g/mol): 563.8
Molecular Formula: C12H6Br2I2
InChi Key: JPXBIAWPJOGFCF-UHFFFAOYSA-N
SMILES: BrC1=CC(I)=C(C=C1)C1=C(I)C=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results