accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOO

CAS RN 852180-60-0

IUPAC Name: 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzaldehyde
Synonyms: 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzaldehyde
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: BJICSBRPQDVEHP-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results