accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 852227-87-3

IUPAC Name: [(1,3-dimethyl-1H-pyrazol-5-yl)methyl](methyl)amine
Synonyms: [(2,5-dimethylpyrazol-3-yl)methyl](methyl)amine
Molecular Weight (g/mol): 139.2
Molecular Formula: C7H13N3
InChi Key: RTTFXSJNRLYKMF-UHFFFAOYSA-N
SMILES: CNCC1=CC(C)=NN1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results