accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 852227-94-2

IUPAC Name: 1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
Synonyms: 1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
Molecular Weight (g/mol): 270.14
Molecular Formula: C15H19BN2O2
InChi Key: GVSCNAOZDQCWJJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)N1C=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results