accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOONH(S)CH3CH3

CAS RN 852288-18-7

IUPAC Name: (2S)-3-(2,2-dimethyl-2H-1,3-benzodioxol-5-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-(2,2-dimethyl-1,3-benzodioxol-5-yl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 459.5
Molecular Formula: C27H25NO6
InChi Key: SHZLOTJPHMTVDI-QFIPXVFZSA-N
SMILES: CC1(C)OC2=C(O1)C=C(C[C@H](NC(=O)OCC1C3=C(C=CC=C3)C3=C1C=CC=C3)C(O)=O)C=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)