accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNO

CAS RN 852431-00-6

IUPAC Name: methyl[(3-phenyl-1,2-oxazol-5-yl)methyl]amine
Synonyms: methyl[(3-phenyl-1,2-oxazol-5-yl)methyl]amine
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: IUBUISGKALKRFN-UHFFFAOYSA-N
SMILES: CNCC1=CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results