accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHHOOF(E/Z)(E/Z)

CAS RN 852475-26-4

IUPAC Name: 3-{4-[3-(3-fluorophenyl)-3-oxoprop-1-en-1-yl]-1-methyl-1H-pyrrol-2-yl}-N-hydroxyprop-2-enamide
Synonyms: 3-{4-[3-(3-fluorophenyl)-3-oxoprop-1-en-1-yl]-1-methylpyrrol-2-yl}-N-hydroxyprop-2-enamide
Molecular Weight (g/mol): 314.32
Molecular Formula: C17H15FN2O3
InChi Key: QRDAPCMJAOQZSU-UHFFFAOYSA-N
SMILES: CN1C=C(C=CC(=O)C2=CC(F)=CC=C2)C=C1C=CC(=O)NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results