accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 852534-22-6

IUPAC Name: N,N-bis(4-bromophenyl)-4-tert-butylaniline
Synonyms: N,N-bis(4-bromophenyl)-4-tert-butylaniline
Molecular Weight (g/mol): 459.23
Molecular Formula: C22H21Br2N
InChi Key: KKLLWAUOZGXWDN-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results