accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NOHN

CAS RN 85264-33-1

IUPAC Name: (3,5-dimethyl-1H-pyrazol-1-yl)methanol
Synonyms: dimethyl hydroxymethyl pyrazole
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: OBENDWOJIFFDLZ-UHFFFAOYSA-N
SMILES: CC1=NN(CO)C(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)