CAS RN 853-35-0
IUPAC Name:
disodium 9,10-dioxo-9,10-dihydroanthracene-1,5-disulfonate
Synonyms:
disodium 9,10-dioxoanthracene-1,5-disulfonate
Molecular Weight (g/mol):
412.29
Molecular Formula:
C14H6Na2O8S2
InChi Key:
OFASSSMJNCWWTP-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC2=C1C(=O)C1=C(C2=O)C(=CC=C1)S([O-])(=O)=O