accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNNOOHNOS

CAS RN 853625-60-2

IUPAC Name: N-{4-[5-(2H-1,3-benzodioxol-5-yl)-3-methoxy-1H-1,2,4-triazol-1-yl]phenyl}-2-(phenylsulfanyl)acetamide
Synonyms: N-{4-[5-(2H-1,3-benzodioxol-5-yl)-3-methoxy-1,2,4-triazol-1-yl]phenyl}-2-(phenylsulfanyl)acetamide
Molecular Weight (g/mol): 460.51
Molecular Formula: C24H20N4O4S
InChi Key: QPGYAMIHXLCFTJ-UHFFFAOYSA-N
SMILES: COC1=NN(C(=N1)C1=CC2=C(OCO2)C=C1)C1=CC=C(NC(=O)CSC2=CC=CC=C2)C=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results