accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 853771-88-7

IUPAC Name: 4-bromo-1-methoxy-2-(trifluoromethoxy)benzene
Synonyms: 4-bromo-1-methoxy-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 271.03
Molecular Formula: C8H6BrF3O2
InChi Key: QVGIROOBNUFIKC-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)(F)F)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results