accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85391-28-2

IUPAC Name: 1-dodecylimidazolidine-2,4-dione
Synonyms: 1-dodecylimidazolidine-2,4-dione
Molecular Weight (g/mol): 268.4
Molecular Formula: C15H28N2O2
InChi Key: IHWNCNSXOZBQBX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN1CC(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results