accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCHZnBr

CAS RN 85459-20-7

IUPAC Name: 1-phenylethanide; bromozincylium
Synonyms: 1-phenylethanide; bromozincylium
Molecular Weight (g/mol): 250.44
Molecular Formula: C8H9BrZn
InChi Key: LCPVHNNFHPWFCI-UHFFFAOYSA-M
SMILES: [Zn+]Br.C[CH-]C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results